Zinkoxid 50% in 1,2-Propandiolmonomethyletheracetat, kolloidale Dispersion mit Detergens, NanoArc® ZN-0625
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Zink(II)oxid
44924.22
44924.22EA
30.3
EUR
44924.22
Zinkoxid 50% in 1,2-Propandiolmonomethyletheracetat, kolloidale Dispersion mit Detergens, NanoArc® ZN-0625
Zinkoxid
CAS No.: 1314-13-2
Molecular Formula: ZnO
Formula Weight: 81.37
Physical Form: 70nm APS for dry powder (typical)
Odour: Ester-like
UN No.: UN1993
MDL No.: MFCD00011300
Applications: Zinc oxide dispersion in Dowanol® PMA suitable for incorporation in a wide variety of coatings formulations
Formel:
ZnO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011300 CAS-Nummer: 1314-13-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...