Tetramethylammoniumhydroxid 1.0 M aqueous solution ACS

Lieferant: Thermo Fisher Scientific
Gefahr

44938.K2 44938.AK
44938.K2EA 192 EUR
44938.K2 44938.AK
Tetramethylammoniumhydroxid 1.0 M aqueous solution ACS
Tetramethylammonium hydroxide

CAS No.: 75-59-2 Molecular Formula: C4H13NO Formula Weight: 91.15 Physical Form: Liquid Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Beilstein Reference: 3558708 Merck Reference: 14,9224 UN No.: 1835 MDL No.: MFCD00008280

Formel: C₄H₁₃NO
Molecular Weight: 91.15 g/mol
Dichte: 1.02 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008280
CAS-Nummer: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8 III
Merck Index: 14,9224

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR