Tetramethylammoniumhydroxid 2,38% w/w aqueous solution , 99,9999% Metals basis, Electronic Grade for the electronics industry

Lieferant: Thermo Fisher Scientific
Gefahr

44940.LU 44940.LV 44940.K2 44940.AK
44940.LUEA 367 EUR
44940.LU 44940.LV 44940.K2 44940.AK
Tetramethylammoniumhydroxid 2,38% w/w aqueous solution , 99,9999% Metals basis, Electronic Grade for the electronics industry
Tetramethylammonium hydroxide

Purity: 99.9999% CAS No.: 75-59-2 Molecular Formula: C4H13NO Formula Weight: 91.15 Physical Form: Liquid Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Beilstein Reference: 3558708 Merck Reference: 14,9224 UN No.: 1835 MDL No.: MFCD00008280

Formel: C₄H₁₃NO
Molecular Weight: 91.15 g/mol
Dichte: 1.02 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008280
CAS-Nummer: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8 III
Merck Index: 14,9224

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR