Iron(II) gluconate hydrate, Supelco®

Lieferant: Merck

Synonyme: Iron(II) Gluconate Hydrate, Iron(II) D-gluconate dihydrate, Gluconic Acid Iron(II) Salt

MQ100

44948-250G
44948-250GEA 91.8 EUR
44948-250G
Iron(II) gluconate hydrate, Supelco®
Iron(II) gluconate hydrate
Formel: C₁₂H₂₄FeO₁₅
Molecular Weight: 464.16 g/mol
MDL: MFCD00150872
CAS-Nummer: 22830-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d472d7c6f93722e7cf70a1313954a160e4702661&vid=d472d7c6f93722e7cf70a1313954a160e4702661&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Loss (on drying) 5 − 15%
Anion traces (chloride)(cl⁻) ≤500 mg/kg
Anion traces (sulphate)(SO₄²⁻) ≤500 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR