Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)

Lieferant: Thermo Fisher Scientific
Achtung

44955.06 44955.14 44955.03
44955.06EA 530 EUR
44955.06 44955.14 44955.03
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)

CAS No.: 16971-33-8 Molecular Formula: C55H46ClOP3Ru Formula Weight: 952.41 Physical Form: Crystalline Sensitivity: Moisture and Light Sensitive Appearance: Gray Melting Point: 204-206° dec. MDL No.: MFCD00049804

Formel: C₅₅H₄₅ClOP₃Ru
Lagertemperatur: Raumtemperatur
MDL: MFCD00049804
CAS-Nummer: 16971-33-8
EINECS: 241-051-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR