Silver conductive ink

Lieferant: Thermo Fisher Scientific
Achtung

44958.14 44958.06
44958.14EA 329 EUR
44958.14 44958.06
Silver conductive ink
Silver conductive ink

Applications: This is a conducting screen printable thick film ink for membrane switch production.


CAS No.: 7440-22-4 Physical Form: Liquid Density: 2.3 Appearance: Silver MDL No.: MFCD01868501

Dichte: 2,3
Lagertemperatur: Raumtemperatur
MDL: MFCD01868501
EINECS: 231-131-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR