Chlorobis(cyclooctene)iridium(I) dimer (∼42,9% Ir)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Chlorobis(cyclooctene)iridium(I)dimer
44966.06
44966.03
44966.06EA
1645
EUR
44966.06
44966.03
Chlorobis(cyclooctene)iridium(I) dimer (∼42,9% Ir)
Chlorobis(cyclooctene)iridium(I) dimer
CAS No.: 12246-51-4
Molecular Formula: C32H56Cl2Ir2
Formula Weight: 896.00
MDL No.: MFCD00213465
Formel:
C₃₂H₅₆Cl₂Ir₂ Molecular Weight: 896.134 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00213465 CAS-Nummer: 12246-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...