Polysorbat 80 (pflanzliche Basis) NF, Multikompendial, raffiniert, J.T.Baker®

Lieferant: Avantor


Veränderungsmanagement-Kategorie= R

4500-04EA 1990 EUR
4500-04 4500-06 4500-03
Polysorbat 80 (pflanzliche Basis) NF, Multikompendial, raffiniert, J.T.Baker®
Polysorbat 80

Polysorbate 80 utilises ingredients of non-animal origin and non-peanut origin.


  • Typical oleic acid content, 77%
  • GMP manufactured product

Schmelzpunkt: –21 °C
Dichte: 1,062 g/cm³
Flammpunkt: 113 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00082107
CAS-Nummer: 9005-65-6
EINECS: 500-019-9

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product ≤2.0
Meets J.P. Chemical Specifications ≤10 ppm
Meets N.F. Requirements 65 - 80
Meets E.P. Chemical Specifications Passes test
CAUTION: For Manufacturing, processing or repackaging Passes test
Bulk Pharmaceutical Chemical ≤0.25%
NF - Acid Value ≤2.0
NF - Hydroxyl Value 65 - 80
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Residue on Ignition ≤0.25 %
NF - Saponification Value 45 - 55
NF - Specific Gravity at 25°C 1.06 - 1.09
NF - Viscosity at 25°C, cSt 300 - 500
NF - Water (H₂O) ≤3.0 %
NF - Peroxide Value ≤10.0
NF - Ethylene Oxide ≤1 ppm
NF - Dioxane ≤10 ppm
NF - Composition of Fatty Acids - Myristic Acid ≤5.0 %
NF - Composition of Fatty Acids - Palmitic Acid ≤16.0 %
NF - Composition of Fatty Acids - Palmitoleic Acid ≤8.0 %
NF - Composition of Fatty Acids - Stearic Acid ≤6.0 %
NF - Composition of Fatty Acids - Oleic Acid ≥58.0 %
NF - Composition of Fatty Acids - Linoleic Acid ≤18.0 %
NF - Composition of Fatty Acids - Linolenic Acid ≤4.0 %
EP - Acid Value ≤2.0
EP - Total Ash ≤0.25 %
EP - Hydroxyl Value 65 - 80
EP - Identification A Passes Test
EP - Identification D Passes Test
EP - Peroxide Value ≤10.0
EP - Ethylene Oxide ≤1 ppm
EP - Dioxan ≤10 ppm
EP - Saponification Value 45 - 55
EP - Water (H₂O) ≤3.0 %
EP/BP - Composition of Fatty Acids - Myristic Acid ≤5.0 %
EP/BP - Composition of Fatty Acids - Palmitic Acid ≤16.0 %
EP/BP - Composition of Fatty Acids - Palmitoleic Acid ≤8.0 %
EP/BP - Composition of Fatty Acids - Stearic Acid ≤6.0 %
EP/BP - Composition of Fatty Acids - Oleic Acid ≥58.0 %
EP/BP - Composition of Fatty Acids - Linoleic Acid ≤18.0 %
EP/BP - Composition of Fatty Acids - Linolenic Acid ≤4.0 %
JP - Appearance Passes Test
JP - Acid Value ≤2.0
JP - Composition of Fatty Acids - Myristic Acid ≤5.0 %
JP - Composition of Fatty Acids - Palmitic Acid ≤16.0 %
JP - Composition of Fatty Acids - Palmitoleic Acid ≤8.0 %
JP - Composition of Fatty Acids - Stearic Acid ≤6.0 %
JP - Composition of Fatty Acids - Oleic Acid ≥58.0 %
JP - Composition of Fatty Acids - Linoleic Acid ≤18.0 %
JP - Composition of Fatty Acids - Linolenic Acid ≤4.0 %
JP - Dioxane ≤10 ppm
JP - Ethylene Oxide ≤1 ppm
JP - Heavy Metals (as Pb) ≤20 ppm
JP - Hydroxyl Value, meq KOH/g 65 - 80
JP - Identification Passes Test
JP - Peroxide Value ≤10.0
JP - Residue on Ignition ≤0.25 %
JP - Water (H₂O) ≤3.0 %
JP - Saponification Value 45 - 55
Additional Tests - Color (APHA) ≤150
Additional Tests - Odor Passes Test
Additional Tests - Water (H₂O) ≤0.2 %
Additional Tests - Peroxide Value (meq/kg) ≤2.0
Additional Tests - Endotoxin Concentration (EU/mL) ≤2
Microbiological - Total Aerobic Microbial Count (cfu/g) ≤100
Microbiological - Escherichia coli Passes Test
Microbiological - Pseudomonas aeruginosa Passes Test
Microbiological - Salmonella Passes Test
Microbiological - Staphylococcus aureus Passes Test
Microbiological - Total Yeast and Mold Count (cfu/g) ≤50
1,4-Dioxane ≤5 ppm
Ethylene glycol ≤ 0.10 %
Diethylene glycol ≤ 0.10 %
Vegetable Based
This product utilizes ingredients of non-animal origin and non-peanut origin.
Suitable for use in the manufacture of parenteral dosage forms.
Only Class 2 (1,4 Dioxane, Ethylene Glycol) and Class 3 (acetic acid, 2-propanol) solvents are likely to be present. Class 2 solvents are below the Option 1 limits and any Class 3 solvent is <0.5%.
Typical Oleic Acid Content, 77%
Metallic Residues: No metal catalysts or metal reagents, as defined by EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of this material.
Due to the anhydrous nature of this product, sodium oleate, a carboxylate salt/soap formed naturally in the process and which can be white to brown in color, can precipitate with time and may affect product viscosity.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR