Essigsäure (Eisessig) 99,8-100,0%, ARISTAR® für die Spurenanalyse
Lieferant: VWR Chemicals
|
Gefahr
|
450013YEA
142
EUR
450013Y
Essigsäure (Eisessig) 99,8-100,0%, ARISTAR® für die Spurenanalyse
Essigsäure
Formel:
H₃CCOOH Siedepunkt: 118 °C (1013 hPa) Schmelzpunkt: 17 °C Dichte: 1,05 g/cm³ (20 °C) Flammpunkt: 38,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036152 CAS-Nummer: 64-19-7 EINECS: 200-580-7 UN: 2789 ADR: 8 (3),II REACH: 01-2119475328-30 |
|
Spezifikation Testergebnisse
Assay | 99.8 - 100.0 % |
Colour | Max. 10 APHA |
Acetaldehyde | Max. 2 ppm |
Acetone | Max. 1 ppm |
Acetic anhydride | Max. 100 ppm |
Freezing point | Min. 16.3 °C |
Cl (Chloride) | Max. 0.1 ppm |
PO4 (Phosphate) | Max. 0.1 ppm |
SO4 (Sulphate) | Max. 0.5 ppm |
Ag (Silver) | Max. 0.010 ppm |
Al (Aluminium) | Max. 0.005 ppm |
As (Arsenic) | Max. 0.003 ppm |
Au (Gold) | Max. 0.020 ppm |
B (Boron) | Max. 0.02 ppm |
Ba (Barium) | Max. 0.005 ppm |
Be (Beryllium) | Max. 0.010 ppm |
Bi (Bismuth) | Max. 0.050 ppm |
Ca (Calcium) | Max. 0.200 ppm |
Cd (Cadmium) | Max. 0.003 ppm |
Cr (Chromium) | Max. 0.010 ppm |
Co (Cobalt) | Max. 0.005 ppm |
Cu (Copper) | Max. 0.003 ppm |
Fe (Iron) | Max. 0.050 ppm |
Ga (Gallium) | Max. 0.010 ppm |
Ge (Germanium) | Max. 0.020 ppm |
Hg (Mercury) | Max. 0.002 ppm |
In (Indium) | Max. 0.010 ppm |
K (Potassium) | Max. 0.050 ppm |
Li (Lithium) | Max. 0.010 ppm |
Mg (Magnesium) | Max. 0.010 ppm |
Mn (Manganese) | Max. 0.005 ppm |
Mo (Molybdenum) | Max. 0.005 ppm |
Na (Sodium) | Max. 0.200 ppm |
Ni (Nickel) | Max. 0.005 ppm |
Pb (Lead) | Max. 0.005 ppm |
Pt (Platinum) | Max. 0.050 ppm |
Sn (Tin) | Max. 0.050 ppm |
Sr (Strontium) | Max. 0.005 ppm |
Ti (Titanium) | Max. 0.050 ppm |
Tl (Thallium) | Max. 0.050 ppm |
V (Vanadium) | Max. 0.010 ppm |
Zn (Zinc) | Max. 0.050 ppm |
Zr (Zirconium) | Max. 0.050 ppm |
Substances reducing K2Cr2O7 | Passes test |
Substances reducing KMnO4 | Passes test |
Dilution test | Passes test |
Non-volatile matter | Max. 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...