Gallium(III)chlorid, ultra trocken ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr


Unter Argon in Ampullen verpackt

45026.22 45026.14 45026.06
45026.22EA 738 EUR
45026.22 45026.14 45026.06
Gallium(III)chlorid, ultra trocken ≥99,999% (Metall-Basis)
Gallium(III)chlorid

Purity: 99.999% CAS No.: 13450-90-3 Molecular Formula: GaCl3 Formula Weight: 176.03 Physical Form: 8mm and down Granules, Ampouled under argon Density: 2.47 Appearance: White Melting Point: 78° Boiling Point: 201° Merck Reference: 14,4348 UN No.: 3260 MDL No.: MFCD00011018

Formel: Cl₃Ga
Siedepunkt: 201 °C
Schmelzpunkt: 78 °C
Dichte: 2,47
Lagertemperatur: Raumtemperatur
MDL: MFCD00011018
CAS-Nummer: 13450-90-3
EINECS: 236-610-0
UN: 3260
ADR: 8,II
Merck Index: 14,4348

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR