Hafnium(IV)fluorid ≥99,9% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

45061.14
45061.14EA 547 EUR
45061.14
Hafnium(IV)fluorid ≥99,9% (Metall-Basis)
Hafnium(IV)fluorid

Purity: 99.9% CAS No.: 13709-52-9 Molecular Formula: HfF4 Formula Weight: 254.48 Physical Form: -60 Mesh Powder Sensitivity: Moisture Sensitive Appearance: White UN No.: 3290 MDL No.: MFCD00016127

Formel: F₄Hf
Molecular Weight: 254,48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016127
CAS-Nummer: 13709-52-9
EINECS: 237-258-0
UN: 3290
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR