Arsen(III)iodid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

45067.18 45067.09
45067.18EA 518 EUR
45067.18 45067.09
Arsen(III)iodid ≥98%
Arsen(III)iodid

Purity: 98% CAS No.: 7784-45-4 Molecular Formula: AsI3 Formula Weight: 455.64 Physical Form: Small Pieces Density: 4.39 Melting Point: 140° Boiling Point: 424° Merck Reference: 14,803 UN No.: 1557 MDL No.: MFCD00014170

Formel: AsI₃
Siedepunkt: 424 °C
Schmelzpunkt: 140 °C
Dichte: 4,39
Lagertemperatur: Raumtemperatur
MDL: MFCD00014170
CAS-Nummer: 7784-45-4
EINECS: 232-068-4
UN: 1557
ADR: 6.1,II
Merck Index: 14,803

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR