Graphit ≥99,997% (Metall-Basis), Stäbe, Ø 10.0 mm (0.40 in)
Lieferant: Thermo Fisher Scientific
45077.DW
45077.DWEA
59.1
EUR
45077.DW
Graphit ≥99,997% (Metall-Basis), Stäbe, Ø 10.0 mm (0.40 in)
Graphit
Purity: 99.997%
Physical Form: UF-4S grade
MDL No.: MFCD00144065
Formel:
C Molecular Weight: 12.01 g/mol Schmelzpunkt: 3827 Dichte: 2.09…2.23 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144065 CAS-Nummer: 7782-42-5 EINECS: 231-955-3 Merck Index: 12,04560 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...