Zinn ≥99,9% (Metall-Basis), Barren/Knopf, Ø ∼36 mm (1.4 in), Dicke 8 mm (0.3 in)
Lieferant: Thermo Fisher Scientific
45110.KF
45110.KFEA
59.9
EUR
45110.KF
Zinn ≥99,9% (Metall-Basis), Barren/Knopf, Ø ∼36 mm (1.4 in), Dicke 8 mm (0.3 in)
Zinn
Purity: 99.9%
CAS No.: 7440-31-5
Physical Form: ≈50g/piece
Appearance: Silver
Odour: Odourless
MDL No.: MFCD00133862
Formel:
Sn Siedepunkt: 2270 °C (1013 hPa) Schmelzpunkt: 232 °C Dichte: 7,31 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00133862 CAS-Nummer: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...