Indium ≥99,99% (Metall-Basis), Barren/Knopf, Ø ∼31 mm (1.2 in), Dicke 5.3 mm (0.21 in)
Lieferant: Thermo Scientific
45111.KF
45111.KFEA
212
EUR
45111.KF
Indium ≥99,99% (Metall-Basis), Barren/Knopf, Ø ∼31 mm (1.2 in), Dicke 5.3 mm (0.21 in)
Indium
Purity: 99.99%
CAS No.: 7440-74-6
Physical Form: ≈28g/piece
Appearance: Silver
Odour: Odourless
MDL No.: MFCD00134048
Formel:
In Molecular Weight: 114,82 g/mol Siedepunkt: 2000 °C (1013 hPa) Schmelzpunkt: 156 °C Dichte: 7,3 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134048 CAS-Nummer: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...