Niob ≥99,95% (Metall-Basis), Barren/Knopf, Ø ∼25.4 mm (1.0 in), Dicke 11 mm (0.45 in)

Lieferant: Thermo Fisher Scientific

45113.KF
45113.KFEA 130 EUR
45113.KF
Niob ≥99,95% (Metall-Basis), Barren/Knopf, Ø ∼25.4 mm (1.0 in), Dicke 11 mm (0.45 in)
Niobium

Purity: 99.95% CAS No.: 7440-03-1 Physical Form: ≈50g/piece Appearance: Silver-grey Odour: Odourless MDL No.: MFCD00011126

Formel: Nb
Molecular Weight: 92.91 g/mol
Siedepunkt: 3700
Schmelzpunkt: 2468
Dichte: 8.57 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011126
CAS-Nummer: 7440-03-1
EINECS: 231-113-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR