Mangan ≥99,9998% (Metall-Basis), Stücke ≤ 10 mm, ultrarein
Lieferant: Thermo Fisher Scientific
45130.14
45130.14EA
1085
EUR
45130.14
Mangan ≥99,9998% (Metall-Basis), Stücke ≤ 10 mm, ultrarein
Manganese
Purity: 99.9998%
CAS No.: 7439-96-5
Molecular Formula: Mn
Physical Form: Ampouled under vacuum
Odour: Odourless
Formel:
Mn Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011111 CAS-Nummer: 7439-96-5 EINECS: 231-105-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...