Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Drahtgeflecht 500 mesh woven from 0.02mm (0.0009in) Type 304
Lieferant: Thermo Scientific
Gefahr
45140.RF
45140.RFEA 506 EUR
45140.RF
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Drahtgeflecht 500 mesh woven from 0.02mm (0.0009in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt% MDL No.: MFCD00148499