Polyethylene, Tafeln, niedrige Dichte, Dicke 3.18 mm (0.125 in)

Lieferant: Thermo Scientific

45175.HB
45175.HBEA 19.8 EUR
45175.HB
Polyethylene, Tafeln, niedrige Dichte, Dicke 3.18 mm (0.125 in)
Polyethylene (CAS 9002-88-4)

CAS No.: 9002-88-4 Physical Form: Sheet Density: 0.92 Appearance: White Odour: Odourless MDL No.: MFCD00084423

Formel: (CH₂CH₂)n
Dichte: 0,92
Lagertemperatur: Raumtemperatur
MDL: MFCD00084423
CAS-Nummer: 9002-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR