Zinc 5,10,15,20-tetra(4-pyridyl)-21H,23H-porphine 90% (Farbstoffgehalt), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
M-Clarity™ Qualitätsstufe = MQ100
452718-1G
452718-1GEA
220
EUR
452718-1G
Zinc 5,10,15,20-tetra(4-pyridyl)-21H,23H-porphine 90% (Farbstoffgehalt), Sigma-Aldrich®
Zinc 5,10,15,20-tetra(4-pyridyl)-21H,23H-porphine
Zinc 5,10,15,20-tetra(4-pyridyl)-21H,23H-porphine (ZnTPyP) forms an organic thin-film, which can be used to fabricate hybrid heterojunction solar cells. It is also used in the production of organic nanostructures, which find applications in organic field-effect transistors (OFET), organic light-emitting displays (OLED), and nanosensors.
Formel:
C₄₀H₂₄N₈Zn Molecular Weight: 682,06 g/mol Schmelzpunkt: >300 °C (Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00274658 CAS-Nummer: 31183-11-6 |
Spezifikation Testergebnisse
Appearance (Colour), Violet or Purple | Conforms to Requirements |
Appearance (Form) | Powder |
Infrared spectrum | Conforms to Structure |
Wavelength, In Chloroform (Ethanol Stabilised) | 420- 426 nm |
Extinction Coefficient | ≥200,000 |
Carbon | 62.3 - 79.2% |
Nitrogen | 14.5 - 18.5% |
Purity (HPLC) | ≥88.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...