Boron 11, 11 B, plasma standard solution, 11 B 100 µg/ml, Specpure®

Lieferant: Thermo Fisher Scientific

45273.AD
45273.ADEA 274 EUR
45273.AD
Boron 11, 11 B, plasma standard solution, 11 B 100 µg/ml, Specpure®
Standards Kationischer Standards Bor-Standards

Molecular Formula: Matrix: H₂O
Physical Form: Liquid
MDL No.: MFCD00151272

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR