Chromium 50, 50 Cr, plasma standard solution, 50 Cr 10 µg/ml, Specpure®
Lieferant: Thermo Fisher Scientific
45275.AD
45275.ADEA
231
EUR
45275.AD
Chromium 50, 50 Cr, plasma standard solution, 50 Cr 10 µg/ml, Specpure®
Standards
Kationischer Standards
Chrom-Standards
Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00190543
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...