1-Iod-3,5-dinitrobenzol 97%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
452769-1G
452769-1GEA
0
EUR
452769-1G
1-Iod-3,5-dinitrobenzol 97%, Sigma-Aldrich®
1-Iod-3,5-dinitrobenzol
Formel:
C₆H₃IN₂O₄ Molecular Weight: 294.01 g/mol Schmelzpunkt: 213...215 °C (Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00024330 CAS-Nummer: 6276-04-6 |
|
Spezifikation Testergebnisse
Appearance (Colour) |
Conforms to Requirements Faint to Dark Yellow |
Appearance (Form) |
Conforms to Requirements Powder or Crystals |
Infrared spectrum | Conforms to Structure |
Carbon | 48.2 - 51.8% |
Nitrogen | 14.0 - 15.1% |
Purity (HPLC) | ≥96.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...