Eisen 57, ⁵⁷Fe, Plasma-Standardlösung, Specpure®

Lieferant: Thermo Fisher Scientific

45277.AD
45277.ADEA 283 EUR
45277.AD
Eisen 57, ⁵⁷Fe, Plasma-Standardlösung, Specpure®
Standards Kationischer Standards Eisen-Standards

UN Nr.: UN3264
MDL Nr.: MFCD00144503


Bitte beachten 10 mg/l = 10 µg/ml=10 ppm

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR