Lithium 6, 6 Li, plasma standard solution, 6 Li 100 µg/ml, Specpure®

Lieferant: Thermo Fisher Scientific

45280.AE
45280.AEEA 250 EUR
45280.AE
Lithium 6, 6 Li, plasma standard solution, 6 Li 100 µg/ml, Specpure®
Standards Kationischer Standards Lithium-Standards

Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00084621

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR