Selenium 78, 78 Se, plasma standard solution, 78 Se 10 µg/ml, Specpure®
Lieferant: Thermo Fisher Scientific
45283.AD
45283.ADEA
141
EUR
45283.AD
Selenium 78, 78 Se, plasma standard solution, 78 Se 10 µg/ml, Specpure®
Standards
Kationischer Standards
Selen-Standards
Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00084579
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...