Lithium nitrate, ionisation buffer solution, Specpure®

Lieferant: Thermo Fisher Scientific

45293.AE
45293.AEEA 102 EUR
45293.AE
Lithium nitrate, ionisation buffer solution, Specpure®
Puffer Ionisationspuffer

CAS No.: 7790-69-4
Molecular Formula: 1% Li (as Li₂CO₃) in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011094

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR