Soxhlet Extraktions-Heizapparat, mehrstellig
Lieferant: GERHARDT
10-0012
453-1997EA 3050 EUR
453-1997
Soxhlet Extraktions-Heizapparat, mehrstellig
Extraktoren Extraktionsapparate, nach Soxhlet
Extraction heating unit for Soxhlet extraction.
  • Temperature individually and continuously variable up to 425 °C
  • Supplied with hot plate top moulds, air bath inserts, support rods (600 × 12 mm) and power cord
  • For round bottom flask from 250 to 500 ml
  • Heater base mould diameter: 85 mm
Supplied without glassware or condenser tubing
Lieferumfang: The cooling water supply system, Soxhlet glass parts and holders for fastening the glass parts to the support rods have to be ordered separately.