Benzamidoxim 97%

Lieferant: Thermo Fisher Scientific
Gefahr

453280010 453280050
453280010.EA 29.6 EUR
453280010. 453280050.
Benzamidoxim 97%
Benzamidoxim
Formel: C₇H₈N₂O
Schmelzpunkt: 74…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00031485
CAS-Nummer: 613-92-3
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point 73 to 79 °C
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR