Supelco® PESTANAL® Organic Reference Standard, Captafol

Lieferant: Merck

45365-250MG
45365-250MGEA 31.9 EUR
45365-250MG
Supelco® PESTANAL® Organic Reference Standard, Captafol
Standards Organische Standards

  • GHS signal: GHS08, GHS07, GHS09


Bitte beachten P201 - P202 - P273 - P280 - P302 + P352 - P308 + P313

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6ea4aa335656179eb7cbb339067b7014f1a1367&vid=b6ea4aa335656179eb7cbb339067b7014f1a1367&prid=web&clid=SAPDAM

Bestellen

TECHNISCHE DATEN

M-Clarity Qualitätsstufe MQ100
Environmentally Preferable
CAS Nr. 2425-06-1
MDL No. MFCD00041816
Molecular weight 349,06
Beschreibung Gefahrenklasse H317 - H350 - H410
Flammpunkt Not applicable
Shipping conditions STD
Lagerbedingungen 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR