2-Methoxyestradiol, Calbiochem®

Lieferant: Merck Millipore Calbiochem

Synonyme: (8R,9S,13S,14S,17S)-2-Methoxy-13-methyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-3,17-diol

454180-10 454180-50
454180-10EA 116 EUR
454180-10 454180-50
2-Methoxyestradiol, Calbiochem®
2-Methoxyestradiol
Formel: C₁₉H₂₆O₃
Molecular Weight: 302.408 g/mol
MDL: MFCD00010489
CAS-Nummer: 362-07-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR