4-(2,4-Dichlorphenoxy)buttersäure (2,4-DB), Supelco®
Lieferant: Merck
45420-250MG-R
45420-250MG-REA
61.9
EUR
45420-250MG-R
4-(2,4-Dichlorphenoxy)buttersäure (2,4-DB), Supelco®
4-(2,4-Dichlorphenoxy)buttersäure (2,4-DB)
Formel:
C₁₀H₁₀Cl₂O₃ |
MDL:
MFCD00002819 CAS-Nummer: 94-82-6 EINECS: 202-366-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...