Dichlofenthion (O-2,4-Dichlorphenyl-O,O-diethylthiophosphat), Supelco®

Lieferant: Merck


MQ100

45432-250MG
45432-250MGEA 61.9 EUR
45432-250MG
Dichlofenthion (O-2,4-Dichlorphenyl-O,O-diethylthiophosphat), Supelco®
Dichlofenthion (O-2,4-Dichlorphenyl-O,O-diethylthiophosphat)
Formel: C₁₀H₁₃Cl₂O₃PS
Molecular Weight: 315.16 g/mol
CAS-Nummer: 97-17-6
EINECS: 202-564-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54523024f8bb701cadd7a08d879f7e438e1c67e1&vid=74004c555beacbe465e0ccb7e74ea286076dc7c2&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR