N-Methoxysuccinyl-Ala-Ala-Pro-Val-p-nitroanilide ≥98%, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
MeOSuc-Ala-Ala-Pro-Val-pNA
454454-50
454454-50EA
157
EUR
454454-50
N-Methoxysuccinyl-Ala-Ala-Pro-Val-p-nitroanilide ≥98%, Millipore®
N-Methoxysuccinyl-Ala-Ala-Pro-Val-p-nitroanilide
Soluble, specific substrate for human leukocyte elastase. It is 200-fold more sensitive than the conventional substrate. Cleavage of substrate can be monitored at ~405 nm.
Formel:
C₂₇H₃₈N₆O₉ Molecular Weight: 590,634 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00077137 CAS-Nummer: 70967-90-7 |
Spezifikation Testergebnisse
Purity | ≥98% by HPLC |
Form | Off-white solid |
Solubility | Soluble in DMF (5 mg/ml), DMSO (5 mg/ml), and ethanol (1 mg/ml) |
Storage | +2 to –8 °C |
Stability | Stable for up to 3 months at –20 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...