Cadmium(II)tellurid ≥99,999% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

Synonyme: Cadmium telluride

45457.14
45457.14EA 183 EUR
45457.14
Cadmium(II)tellurid ≥99,999% (Metall-Basis)
Cadmium(II)tellurid

Purity: 99.999% CAS No.: 1306-25-8 Molecular Formula: CdTe Formula Weight: 240.00 Physical Form: 6mm & down polycrystalline pieces Density: 6.20 Appearance: Black Odour: Odourless Melting Point: 1041° Merck Reference: 14,1629 UN No.: 3077 MDL No.: MFCD00015998

Formel: CdTe
Molecular Weight: 240,01 g/mol
Schmelzpunkt: 1041 °C
Dichte: 6,20
Lagertemperatur: Raumtemperatur
MDL: MFCD00015998
CAS-Nummer: 1306-25-8
EINECS: 215-149-9
UN: 3077
ADR: 9,III
Merck Index: 14,1629

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR