4-tert-Butylphthalsäureanhydrid 95%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 5-(tert-Butyl)isobenzofuran-1,3-dione

454648-5G
454648-5GEA 225 EUR
454648-5G
4-tert-Butylphthalsäureanhydrid 95%, Sigma-Aldrich®
4-tert-Butylphthalsäureanhydrid

4-tert-Butylphthalic anhydride has been used to synthesise novel yellow quinophthalone dyes for LCD colour filters.

Formel: C₁₂H₁₂O₃
Molecular Weight: 204.2219 g/mol
Schmelzpunkt: 70...75 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00060127
CAS-Nummer: 32703-79-0
EINECS: 251-165-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23d3d6e8a13e949508c9945de60843261397622e&vid=23d3d6e8a13e949508c9945de60843261397622e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white powder, crystals, Crystalline powder and/or chunks
Infrared spectrum Conforms to structure
Titration 94.0%-106.0% (with morpholine)
Gas liquid 94.0% (minimum)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR