2,6-Diphenyl-4H-thiopyran-4-one ≥96%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

454656-500MG
454656-500MGEA 268 EUR
454656-500MG
2,6-Diphenyl-4H-thiopyran-4-one ≥96%, Sigma-Aldrich®
2,6-Diphenyl-4H-thiopyran-4-one

2,6-Diphenyl-4H-thiopyran-4-one,96%

Formel: C₁₇H₁₂OS
Molecular Weight: 264,34 g/mol
Schmelzpunkt: 126…130 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00799527
CAS-Nummer: 1029-96-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c18be94b6bf9c440bb0896c282e041e47ca77ee&vid=f8c85a24b5b64ee70a71036cc2606fb0732a91e4&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR