Nickelsulfid ≥99% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Nickel subsulphide, Nickel disulphane

45498.06
45498.06EA 211 EUR
45498.06
Nickelsulfid ≥99% (Metall-Basis)
Nickelsulfid

Purity: 99% CAS No.: 12035-51-7 Molecular Formula: NiS1.4 Physical Form: Powder UN No.: 3077 MDL No.: MFCD00016266

Formel: Ni₃S₂
Molecular Weight: 240,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00217044
CAS-Nummer: 12035-72-2
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR