Mangan ≥99,9% (Metall-Basis), Nano-Pulver APS 30-50 nm

Lieferant: Thermo Fisher Scientific
Achtung

45508.14
45508.14EA 1010 EUR
45508.14
Mangan ≥99,9% (Metall-Basis), Nano-Pulver APS 30-50 nm
Manganese

Purity: 99.9% CAS No.: 7439-96-5 Molecular Formula: Mn(core)/Mn3O4(shell) Physical Form: Mn3O4 layer 5-12nm UN No.: 3089 MDL No.: MFCD00011111

Formel: Mn
Molecular Weight: 54.94 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011111
CAS-Nummer: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR