Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) (15% Pd)

Lieferant: Thermo Fisher Scientific
Achtung

45511.03
45511.03EA 174 EUR
45511.03
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) (15% Pd)
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II)

CAS No.: 887919-35-9 Molecular Formula: C32H56Cl2N2P2Pd Formula Weight: 708.08 Physical Form: Crystalline Appearance: Yellow MDL No.: MFCD09265123

Formel: C₃₂H₅₆Cl₂N₂P₂Pd
Lagertemperatur: Raumtemperatur
MDL: MFCD09265123
CAS-Nummer: 887919-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR