tri-Natriumcitrat, wasserfrei 99% (durch titrimetrische Analyse)

Lieferant: Thermo Fisher Scientific

45556.A1 45556.30 45556.A7
45556.A1EA 114 EUR
45556.A1 45556.30 45556.A7
tri-Natriumcitrat, wasserfrei 99% (durch titrimetrische Analyse)
tri-Natriumcitrat

Trisodium citrate is used in food additives, beverages, instant drinks, pharmaceuticals and detergents. It is also used as buffering agent, acidity regulator, sequestering or emulsifying agent.


It is an effective agent for removal of carbonate scale from boilers and for cleaning automobile radiators. In the medical field, it is used as an osmotic laxative and an active component of the WHO oral rehydration solution. It is also employed to control acidity in some substances such as gelatin desserts. Further, it is used as a flavour or as a preservative. In addition to this, it is used for the preservation of blood in blood banks.

Formel: C₆H₅Na₃O₇
Schmelzpunkt: 300 °C
Dichte: 1,815 g/cm³ (20 °C)
Flammpunkt: 290 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012462
CAS-Nummer: 68-04-2
EINECS: 200-675-3
Merck Index: 14,8602

Bestellen

Spezifikation Testergebnisse

Assay (Titration) >98.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR