Calciumthioglycolat Trihydrat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Calciumbis(mercaptoacetat) Trihydrat
45557.18
45557.18EA
131
EUR
45557.18
Calciumthioglycolat Trihydrat ≥98%
Calciumthioglycolat Trihydrat
Purity: 98%
CAS No.: 65208-41-5
Molecular Formula: C4H6CaO4S2.3H2O
Formula Weight: 276.34 (222.29anhy)
Physical Form: Powder
Appearance: White
Melting Point: 280-290° dec.
Beilstein Reference: 3755791
Merck Reference: 14,1711
MDL No.: MFCD00150707
Formel:
C₄H₆CaO₄S₂·3H₂O Molecular Weight: 276,35 g/mol Schmelzpunkt: 280…290 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150707 CAS-Nummer: 65208-41-5 EINECS: 212-402-5 Merck Index: 14,1711 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...