Indium(III)sulfid ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Diindiumtrisulfid, Indiumtrisulfid

45563.04 45563.09 45563.18
45563.04EA 97 EUR
45563.04 45563.09 45563.18
Indium(III)sulfid ≥99,999% (Metall-Basis)
Indium(III)sulfid

Purity: 99.999% CAS No.: 12030-24-9 Molecular Formula: In2S3 Formula Weight: 325.83 Physical Form: Powder Sensitivity: Moisture Sensitive MDL No.: MFCD00011061

Formel: In₂S₃
Molecular Weight: 325.84 g/mol
Schmelzpunkt: 1050 °C
Dichte: 4.89 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011061
CAS-Nummer: 12030-24-9
EINECS: 234-742-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR