Monolinuron (N-(4-Chlorphenyl)-N'-methoxy,N'-methylharnstoff), Supelco®

Lieferant: Merck

Synonyme: 3-(4-Chlorophenyl)-1-methoxy-1-methylurea, Monolinuron

M-Clarity™ Qualitätsstufe = MQ100

45590-250MG
45590-250MGEA 60.2 EUR
45590-250MG
Monolinuron (N-(4-Chlorphenyl)-N'-methoxy,N'-methylharnstoff), Supelco®
Monolinuron (N-(4-Chlorphenyl)-N'-methoxy,N'-methylharnstoff)
Formel: C₉H₁₁ClN₂O₂
MDL: MFCD00055264
CAS-Nummer: 1746-81-2
EINECS: 217-129-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d269601803416ec84867e0895359306cf04b5b5&vid=8575f0bceda2553c85f742e125045d2f992aa5b7&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR