Hexane (mixture of isomers) 99+% for HPLC

Lieferant: Thermo Fisher Scientific
Gefahr

45652.K2 45652.K7
45652.K2EA 72.7 EUR
45652.K2 45652.K7
Hexane (mixture of isomers) 99+% for HPLC
Hexan (Isomerengemisch)
Formel: C₆H₁₄
Siedepunkt: 65…70 °C
Schmelzpunkt: –95 °C
Dichte: 0,67
Flammpunkt: <–23 °C (–9 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009406
CAS-Nummer: 92112-69-1
EINECS: 295-570-2
UN: 1208
ADR: 3,II
Merck Index: 13,04712

Bestellen

Spezifikation Testergebnisse

Optical Absorbance at 195nm ≤1.00AU
Optical Absorbance at 225nm ≤0.050AU
Optical Absorbance at 250nm ≤0.010AU
Optical Absorbance at 275nm ≤0.005AU
Comment Submicron filtered
Impurity content (Heptachlorepoxide by GC-ECD responsive residue) ≤10 ppt ( single peak)
Water Content (Karl Fischer Titration) ≤0.01%
Optical Absorbance at 300nm ≤0.005AU
Residue on Evaporation ≤5 mg/ml
Assay (GC) ≤99%, as C6 Hydrocarbons


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR