Methylenblau, Millipore®
Lieferant: Merck Millipore Calbiochem
|
Achtung
|
Synonyme:
3,7-Bis(dimethylamino)phenothiaziniumchlorid, N,N,N′,N′-Tetramethylthioninchlorid, Methylthioniniumchlorid
457250-1
457250-1EA
46.9
EUR
457250-1
Methylenblau, Millipore®
Methylenblau
Formel:
C₁₆H₁₈ClN₃S Molecular Weight: 319,86 g/mol Schmelzpunkt: 180 °C Dichte: 1,757 g/cm³ (22 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150008 CAS-Nummer: 61-73-4 EINECS: 200-515-2 Merck Index: 12,06137 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...