Gadolinium, Aufdampfplatte, Ø 50.8 mm, Dicke 3.18 mm
Lieferant: Thermo Scientific
45737.KS
45737.KSEA
992
EUR
45737.KS
Gadolinium, Aufdampfplatte, Ø 50.8 mm, Dicke 3.18 mm
Gadolinium
CAS No.: 7440-54-2
MDL No.: MFCD00011022
Formel:
Gd Molecular Weight: 157,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011022 CAS-Nummer: 7440-54-2 EINECS: 231-162-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...