Pyromellitsäuredianhydrid (1,2,4,5-Benzoltetracarbonsäuredianhydrid) ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98%
CAS No.: 89-32-7
Molecular Formula: C10H2O6
Formula Weight: 218.12
Flash Point: 380°(716°F)
Physical Form: Crystalline powder
Density: 1.68
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Melting Point: 284-288°
Boiling Point: 305-310°/30mm
Beilstein Reference: 213583
MDL No.: MFCD00005005
Formel:
C₁₀H₂O₆ Siedepunkt: 305…310 °C (30 mmHg) Schmelzpunkt: 284…288 °C Dichte: 1,68 Flammpunkt: 380 °C (716 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005005 CAS-Nummer: 89-32-7 EINECS: 201-898-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...