Crucibles, quartz, self sealing
Lieferant: CEM
S-322200
458-4221EA 3750 EUR
458-4221
Crucibles, quartz, self sealing
Tiegel
This quartz crucible is self sealing and enables oxygen free ashing.
Lieferumfang: Includes crucible and lid.