Automated peptide synthesis, liberty
Lieferant: CEM
925437
458-4254EA 0 EUR
458-4254
Automated peptide synthesis, liberty
Verdampfer Parallelsynthese-Systeme
Make higher purity peptides faster. Microwave energy drives biochemical reactions to completion up to 10 times faster than by conventional methods, resulting in peptides of greater purity than can be produced under conventional methods. Reactions are not just faster; they are more complete when driven by microwave energy.
  • Greater purity peptides than conventional synthesis
  • Totally automated synthesis of up to 12 peptides
  • Wide scale range of 0,025 to 5 mmol
  • Unique resin transfer process allows for a single reaction vessel in a less complicated system
  • Programmable cleavage & automated resin addition
Use either 125- or 250-ml amino acid bottles.
All parts that come into contact with chemicals are composed of chemically-inert materials.